Manual detergent, Emerald
Lieferant: TRISTEL SOLUTIONS
CRY309 CRY310 CRY307
TRSTCRY309EA 1320 EUR
TRSTCRY309 TRSTCRY310 141-0944
Manual detergent, Emerald
Reinigungsmittel Waschmittel und Oberflächenreiniger
A high foam, neutral, detergent for manual cleaning.
  • Neutral detergent for light to medium soiling
  • Dilution for use: Between 0,5% to 2% v/v
A manual detergent for the removal of light to medium soiling. Areas of use include walls, floors, surfaces, ceilings, equipment and utensils. Emerald is a soft metal safe detergent suitable for use on a variety of surfaces including plastics, silicone, vinyl and aluminium.
The crystel detergents are specifically formulated for the removal of the various residues that are present in the manufacturing and production process within pharmaceutical and personal care industries.
All detergents are supported by comprehensive technical files which provide all the documentation for validation, including residue test methods.
All detergents are supported by comprehensive technical files which provide all the documentation for validation, including residue test methods.
Lieferumfang: Supplied with its own certificate of analysis. A detailed technical file (including: product specification, manufacturing process, shelf life, microbiological and analytical data) is available on request.
Verpackung: Available in 5, 20, 200 and 1000 litre pack sizes.