Halogen lamp
Lieferant: TECHNICAL SERVICE SPARE PARTS
55147
TSSP55147EA 118 EUR
TSSP55147
Halogen lamp
Beleuchtung Halogenleuchten
Narva halogen special lamps are specifically designed for special lighting tasks. These lamps with prefocus base are suitable for scientific-technical equipment due to their design and optical precision.
  • Made of quartz glass
  • Transmits UV radiation
  • Operating position of s90
  • With prefocus base and mounting plate
Applications: Microscopy, ophthalmology, dermatology, food analysis, blood analysis, illumination for surgeries and analysis, measurement technology, projection.