Glassware Washer for Laboratories, TIVA8
Lieferant: TUTTNAUER
ATS238-0493
TUTTATS238-0493EA 0 EUR
TUTTATS238-0493
Glassware Washer for Laboratories, TIVA8
Reinigungs- und Desinfektionsautomaten
Tuttnauer's TIVA8 Lab line Washers contain unrivalled levels of efficiency that deliver on water and energy
savings. The high-quality system is specifically designed for research intensive laboratory settings that offer
automated cleaning and drying of typical laboratory glassware, as well as miscellaneous plastic items used in
research facilities.
  • Simple operation and process monitoring
  • Advanced control and warning system
  • Easy to clean and maintain process monitoring
  • Optimal results
  • Highest design standards
A wide range of racks, nozzles and accessories complete these machines, suitable to wash and dry a wide variety of laboratory glassware. Door resistant to aggressive chemicals and ergonomic handle durable stainless steel door.
Max pump flow rate: 370 L/min
Max drying fan flow rate with filter: 180 m³/h
Max absorbed power*: 6,2 kW
Max number of dosing pumps: 4
Chemical tank capacity: 5 L