Safety goggles, 6X1, perfect hybrid
Lieferant: UNIVET
6X1.00.00.00
UNVT6X1.00.00.00EA 149 EUR
UNVT6X1.00.00.00
Safety goggles, 6X1, perfect hybrid
Vollsichtbrillen
6X1 combines the lightness of a spectacle to the benefits of a mask. The adjustable temples with SoftPad technology can be easily replaced with the elastic band included.
  • Gun metal frame
  • Wide lens for a field of view without distortion
  • Temples adjustable in length and inclination with SoftPad technology included for absolute versatility
  • Sophisticated indirect ventilation system
  • Soft internal rubber for comfort without local pressure
Provides protection against drops, sprays and large dust particles guaranteed by the special design of the internal rubber.
Weight: 54 g
Standards: EN 166-3; EN 166-4
Lieferumfang: Available with RX insert for corrective lenses.