Tubing cutter for fused silica tubing, Upchurch Scientific®
Lieferant: UPCHURCH SCIENTIFIC
FS-315
UPCHFS-315EA 1560 EUR
UPCHFS-315
Tubing cutter for fused silica tubing, Upchurch Scientific®
Schneidewerkzeuge
Precision cutter for fused silica tubing ensures clean, trouble free cutting of fused silica tubing.
It also includes a built-in magnifying glass to examine the cut tubing ends.