Micro injection port adapter assembly, Upchurch Scientific®
Lieferant: UPCHURCH SCIENTIFIC
M-432-03
UPCHM-432-03EA 125 EUR
UPCHM-432-03
Micro injection port adapter assembly, Upchurch Scientific®
Fittings und Verbinder
This micro injection port adapter assembly can be connected to 360 µm Ext.Ø capillary tubing to introduce samples. It accepts standard 22 gauge Hamilton-style injection syringe needles. The needle butts directly against the connecting tubing during injections, therefore no additional swept volume is added to the fluid pathway by this adapter.
  • For 360 µm Ext.Ø. tubing
  • Mount on bracket or bulkhead