Vorsäulen- und Inline-MicroFilter, Upchurch Scientific®
Lieferant: UPCHURCH SCIENTIFIC
A-735X M-555-1 M-560-1
UPCHA-735XEA 257 EUR
UPCHA-735X UPCHM-555-1 554-3086
Vorsäulen- und Inline-MicroFilter, Upchurch Scientific®
Filter Vorsäulen-Filter und -Fritten
Precolumn MicroFilters are specifically designed to directly connect into microbore or analytical columns.
Total theoretical void volume is only 0,5 μl (includes frit volume) and the PEEK tubing used in the assembly of these units has a 125 μm int.Ø, virtually eliminating any mixing of the sample with the mobile phase.