Upchurch Scientific® Super Flangeless™, P-259x, ferrule, ETFE yellow/stainless steel, 1/4-28 or M6 flat bottom port, for 1/16" OD tubing, 1350 psi
Lieferant: UPCHURCH SCIENTIFIC
Upchurch Scientific® Super Flangeless™, P-259x, ferrule, ETFE yellow/stainless steel, 1/4-28 or M6 flat bottom port, for 1/16" OD tubing, 1350 psi