Forskolin

Lieferant: UPSTATE TECHNOLOGY

Synonyme: (3R,4aR,5S,6S,6aS,10S,10aR,10bS)-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-3-vinyldodecahydro-1H-benzo[f]chromen-5-yl acetate

19-140
UPST19-140EA 0 EUR
UPST19-140
Forskolin
Forskolin
Formel: C₂₂H₃₄O₇
Molecular Weight: 410,51 g/mol
MDL: MFCD00082317
CAS-Nummer: 66575-29-9
EINECS: 266-410-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR