5-Brom-2'-deoxyuridin (Broxuridin)

Lieferant: UPSTATE TECHNOLOGY

Synonyme: 5-Bromo-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 5-Bromo-2'-deoxyuridine, 5-BrdU

19-160
UPST19-160EA 0 EUR
UPST19-160
5-Brom-2'-deoxyuridin (Broxuridin)
5-Brom-2'-deoxyuridin (Broxuridin)
Formel: C₉H₁₁BrN₂O₅
Molecular Weight: 307,10 g/mol
MDL: MFCD00006529
CAS-Nummer: 59-14-3
EINECS: 200-415-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR