Resveratrol

Lieferant: UPSTATE TECHNOLOGY

Synonyme: trans-3,4,5-Trihydroxystilbene, (E)-5-(4-Hydroxystyryl)benzene-1,3-diol

19-172
UPST19-172EA 0 EUR
UPST19-172
Resveratrol
Resveratrol
Formel: C₁₄H₁₂O₃
Molecular Weight: 228,25 g/mol
Schmelzpunkt: 260 °C
Dichte: 1,36 g/cm³ (20 °C)
Lagertemperatur: Gefrierschrank
MDL: MFCD00133799
CAS-Nummer: 501-36-0
Merck Index: 12,08326

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR