di-Natrium-4-nitrophenylphosphat Hexahydrat (pNPP), Tabletten

Lieferant: UPSTATE TECHNOLOGY

20-106
UPST20-106EA 0 EUR
UPST20-106
di-Natrium-4-nitrophenylphosphat Hexahydrat (pNPP), Tabletten
di-Natrium-4-nitrophenylphosphat Hexahydrat (pNPP)
Formel: C₆H₄NNa₂O₆P·6H₂O
Schmelzpunkt: 300 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00066288
CAS-Nummer: 4264-83-9
EINECS: 224-246-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR