(Benzotriazol-1-yloxy)tris(dimethylamino)phosphoniumhexafluorophosphat (BOP)

Lieferant: UPSTATE TECHNOLOGY

Synonyme: 1H-Benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate, ((1H-Benzo[d][1,2,3]triazol-1-yl)oxy)tris(dimethylamino)phosphonium hexafluorophosphate(V), BOP reagent, Benzotriazol-1-yloxytris(dimethylamino)-phosphonium hexafluorophosphate

20-173
UPST20-173EA 0 EUR
UPST20-173
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphoniumhexafluorophosphat (BOP)
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphoniumhexafluorophosphat (BOP)
Formel: C₁₂H₂₂F₆N₆OP₂
Molecular Weight: 442.28 g/mol
MDL: MFCD00011948
CAS-Nummer: 56602-33-6
EINECS: 260-279-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR