Sand
Lieferant: Uptima
Synonyme:
Silicium(IV)oxid, Siliciumdioxid
75035A
UPTI75035AEA
0
EUR
UPTI75035A
Sand
Sand
Formel:
SiO₂ Molecular Weight: 60.08 g/mol Schmelzpunkt: ∼1700 °C Dichte: ∼2.6 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD02100519 CAS-Nummer: 7631-86-9 EINECS: 231-545-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...