2,6-Difluor-4-hydroxybenzonitril

Lieferant: Uptima

Synonyme: 4-Cyano-3,5-difluorophenol

BC111
UPTIBC111EA 0 EUR
UPTIBC111
2,6-Difluor-4-hydroxybenzonitril
2,6-Difluor-4-hydroxybenzonitril
Formel: C₇H₃F₂NO
Molecular Weight: 155.1 g/mol
MDL: MFCD03094499
CAS-Nummer: 123843-57-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR