Benzophenon-4-maleimid

Lieferant: Uptima

Synonyme: 1-(4-Benzoylphenyl)-1H-pyrrole-2,5-dione, 4-(N-Maleimido)benzophenone, Benzophenone-4-maleimide

BI1331
UPTIBI1331EA 0 EUR
UPTIBI1331
Benzophenon-4-maleimid
Benzophenon-4-maleimid
Formel: C₁₇H₁₁NO₃
Molecular Weight: 277.2700 g/mol
MDL: MFCD00079465
CAS-Nummer: 92944-71-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR