4-(Iodacetyl)benzophenon

Lieferant: Uptima

Synonyme: 4-(N-Iodoacetamide)benzophenone, N-(4-Benzoylphenyl)-2-iodoacetamide, Benzophenone-4-Iodoacetamide

BI1351
UPTIBI1351EA 0 EUR
UPTIBI1351
4-(Iodacetyl)benzophenon
4-(Iodacetyl)benzophenon
Formel: C₁₅H₁₁IO₂
Molecular Weight: 365.17 g/mol
CAS-Nummer: 76809-63-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR