4-(Succinimidylcarboxy)benzophenon

Lieferant: Uptima

Synonyme: 4-Benzoylbenzoic acid N-succinimidyl ester, 2,5-Dioxopyrrolidin-1-yl 4-benzoylbenzoate

BI1361
UPTIBI1361EA 0 EUR
UPTIBI1361
4-(Succinimidylcarboxy)benzophenon
4-(Succinimidylcarboxy)benzophenon
Formel: C₁₈H₁₃NO₄
Molecular Weight: 323.2995 g/mol
MDL: MFCD00058572
CAS-Nummer: 91990-88-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR