1-Hexyl-3-methylimidazoliumbromid

Lieferant: Uptima

CH577
UPTICH577EA 0 EUR
UPTICH577
1-Hexyl-3-methylimidazoliumbromid
1-Hexyl-3-methylimidazolium bromide
Formel: C₁₀H₁₉BrN₂
Molecular Weight: 247,18 g/mol
MDL: MFCD03427612
CAS-Nummer: 85100-78-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR