Zinknitrid

Lieferant: Uptima

IN9305
UPTIIN9305EA 0 EUR
UPTIIN9305
Zinknitrid
Zinknitrid
Formel: N₂Zn₃
Molecular Weight: 224,12 g/mol
MDL: MFCD00061327
CAS-Nummer: 1313-49-1
EINECS: 215-207-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR