5-Brom-6-chlor-3-indolylphosphat p-Toluidinsalz (BCIP-Rot)
Lieferant: Uptima
Synonyme:
BCIP Red, Magenta™ phosphate
M1636A
UPTIM1636AEA
0
EUR
UPTIM1636A
5-Brom-6-chlor-3-indolylphosphat p-Toluidinsalz (BCIP-Rot)
5-Brom-6-chlor-3-indolylphosphat p-Toluidinsalz (BCIP-Rot)
Formel:
C₈H₆BrClNO₄P·C₇H₉N |
MDL:
MFCD00210023 CAS-Nummer: 6769-80-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...