α-Cyclodextrin Hydrat

Lieferant: Uptima

U03449
UPTIU03449EA 0 EUR
UPTIU03449
α-Cyclodextrin Hydrat
α-Cyclodextrin Hydrat
Formel: C₃₆H₆₀O₃₀·xH₂O
MDL: MFCD00150804
CAS-Nummer: 51211-51-9
EINECS: 233-007-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR