Glycin (H-Gly-OH)
Lieferant: Uptima
UP018225
UPTIUP018225EA
0
EUR
UPTIUP018225
Glycin (H-Gly-OH)
Glycin (H-Gly-OH)
Formel:
NH₂CH₂COOH Siedepunkt: 181 °C (1013 hPa) Schmelzpunkt: 83 °C Dichte: 1,64 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008131 CAS-Nummer: 56-40-6 EINECS: 200-272-2 Merck Index: 13,04504 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...