N-Hydroxysuccinimid

Lieferant: Uptima

Synonyme: 1-Hydroxyazacyclopentan-2,5-dion, 1-Hydroxy-1,4-butandisäureimid, NHS, 1-Hydroxybernsteinsäureimid, 1-Hydroxy-2,5-pyrrolidindion, HOSu, N-Hydroxybernsteinsäureimid, N-Hydroxy-1,4-butandisäureimid

UP04594A
UPTIUP04594AEA 0 EUR
UPTIUP04594A
N-Hydroxysuccinimid
N-Hydroxysuccinimid
Formel: C₄H₅NO₃
Molecular Weight: 115,09 g/mol
Siedepunkt: 363 °C (1013 hPa)
Schmelzpunkt: 93…95 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00005516
CAS-Nummer: 6066-82-6
EINECS: 228-001-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR