2-(4-Hydroxyphenylazo)benzoesäure

Lieferant: Uptima

Synonyme: 4-Hydroxyazobenzene-2'-carboxylic acid, 2-(4-Hydroxyphenylazo)benzoic acid, 4'-Hydroxyazobenzene-2-carboxylic acid, 2-((4-Hydroxyphenyl)diazenyl)benzoic acid, HABA

UP05361D
UPTIUP05361DEA 0 EUR
UPTIUP05361D
2-(4-Hydroxyphenylazo)benzoesäure
2-(4-Hydroxyphenylazo)benzoesäure
Formel: C₁₃H₁₀N₂O₃
Molecular Weight: 242,23 g/mol
MDL: MFCD00002428
CAS-Nummer: 1634-82-8
EINECS: 216-655-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR