3,3',5,5'-Tetramethylbenzidin (TMB), Pulver, ultrarein
Lieferant: Uptima
UP15426D
UPTIUP15426DEA
0
EUR
UPTIUP15426D
3,3',5,5'-Tetramethylbenzidin (TMB), Pulver, ultrarein
3,3',5,5'-Tetramethylbenzidin (TMB)
Formel:
C₁₆H₂₀N₂ Siedepunkt: 402 °C (1013 hPa) Schmelzpunkt: 166…170 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007748 CAS-Nummer: 54827-17-7 EINECS: 259-364-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...