N-Octyl-β-D-glucopyranosid

Lieferant: Uptima
Achtung

Synonyme: Octyl-β-D-glucosid, Octyl-β-D-glucopyranosid

UP263700 UP263701
UPTIUP263700EA 0 EUR
UPTIUP263700 UPTIUP263701
N-Octyl-β-D-glucopyranosid
N-Octyl-β-D-glucopyranosid
Formel: C₁₄H₂₈O₆
Molecular Weight: 292.37 g/mol
Schmelzpunkt: 110
Lagertemperatur: Gefrierschrank
MDL: MFCD00063288
CAS-Nummer: 29836-26-8
EINECS: 249-887-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR