4-Methylumbelliferylphosphat Dinatriumsalz (4-MUP-Na2)

Lieferant: Uptima

Synonyme: Disodium 4-methylumbelliferyl phosphate, 4-MUP

UP30045A
UPTIUP30045AEA 0 EUR
UPTIUP30045A
4-Methylumbelliferylphosphat Dinatriumsalz (4-MUP-Na2)
4-Methylumbelliferylphosphat Dinatriumsalz (4-MUP-Na2)
Formel: C₁₀H₇Na₂O₆P
Molecular Weight: 300.11 g/mol
MDL: MFCD00037614
CAS-Nummer: 22919-26-2
EINECS: 245-325-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR