Brij® 58

Lieferant: Uptima

Synonyme: Polyoxyethylene (20) cetyl ether, Polyethylene glycol hexadecyl ether

UP314740 UP314741
UPTIUP314740EA 0 EUR
UPTIUP314740 UPTIUP314741
Brij® 58
Brij® 58
Formel: C₅₆H₁₁₄O₂₁
Molecular Weight: 1123.51 g/mol
MDL: MFCD00080892
CAS-Nummer: 9004-95-9
EINECS: 500-014-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR