X-Gal (5-Brom-4-chlor-3-indolyl-ß-D-galactopyranosid)

Lieferant: Uptima

UP40534M UP40534N
UPTIUP40534MEA 0 EUR
UPTIUP40534M UPTIUP40534N
X-Gal (5-Brom-4-chlor-3-indolyl-ß-D-galactopyranosid)
X-Gal (5-Brom-4-chlor-3-indolyl-ß-D-galactopyranosid)
Formel: C₁₄H₁₅BrClNO₆
Schmelzpunkt: 237…239 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00005666
CAS-Nummer: 7240-90-6
EINECS: 230-640-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR