Octyl-β-D-thioglucopyranosid
Lieferant: Uptima
Synonyme:
Octyl-thioglucoside, OTG, 1-S-Octyl-β-D-thioglucopyranoside, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(octylthio)tetrahydro-2H-pyran-3,4,5-triol, N-Octyl-1-thio-Beta-D-glucopyranoside, Octyl β-D-thioglucopyranoside
UP602080
UP602081
UPTIUP602080EA
0
EUR
UPTIUP602080
UPTIUP602081
Octyl-β-D-thioglucopyranosid
Octyl-β-D-thioglucopyranosid
Formel:
C₁₄H₂₈O₅S Molecular Weight: 308.44 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00012189 CAS-Nummer: 85618-21-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...