TRITON™ X-305 (Polyethylenglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]ether, Polyethylenglycol-tert-octylphenylether)

Lieferant: Uptima
Gefahr

UP708534 UP708535
UPTIUP708534EA 0 EUR
UPTIUP708534 UPTIUP708535
TRITON™ X-305 (Polyethylenglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]ether, Polyethylenglycol-tert-octylphenylether)
TRITON™ X-305 (Polyethylenglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]ether, Polyethylenglycol-tert-octylphenylether)
Lagertemperatur: Raumtemperatur
CAS-Nummer: 9002-93-1
UN: 3082
ADR: 9,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR