Ethidiumbromid 10 mg/ml aqueous solution

Lieferant: Uptima
Gefahr

Synonyme: 3,8-Diamino-5-ethyl-6-phenylphenanthridiniumbromid

UP89244B
UPTIUP89244BEA 0 EUR
UPTIUP89244B
Ethidiumbromid 10 mg/ml aqueous solution
Ethidiumbromid
Formel: C₂₁H₂₀BrN₃
Molecular Weight: 394.31 g/mol
Siedepunkt: 72
MDL: MFCD00011724
CAS-Nummer: 1239-45-8
EINECS: 214-984-6
Merck Index: 13,04751

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR