Tripod stands
Lieferant: USBECK Laborgeräte
2031 2033
USBE2031EA 31.1 EUR
USBE2031 USBE2033
Tripod stands
Stative
Retort stand base in tripod shape made from powder-coated steel.
  • For rods up to 13 mm Ø
  • With hinged screw M 6
Thickness: 10 mm