Clamps, flexible, with bosshead, swivel type
Lieferant: USBECK Laborgeräte
2680 2682
USBE2680EA 38.5 EUR
USBE2680 USBE2682
Clamps, flexible, with bosshead, swivel type
Klemmen Klemmen für den allgemeinen Gebrauch
Flexible clamps with bosshead (swivel type). Made of zinc die-cast and brass, nickel plated.
  • With vinyl coated jaws
  • Three-pronged or round jaws
Length: 300 mm