Spoon, Open
Lieferant: USBECK Laborgeräte
3408
USBE3408EA 4.3 EUR
USBE3408
Spoon, Open
Löffel
Spoon with open end made from stainless steel.
  • Autoclavable
  • Easy to clean
  • 25 mm width
  • 29 g weight
Excellent chemical resistance. High temperature resistance.