Gauzes with Stainless Steel Centre
Lieferant: USBECK Laborgeräte
5072
USBE5072EA 5 EUR
USBE5072
Gauzes with Stainless Steel Centre
Draht
Mesh from galvanised steel wire.
  • Alternative to gauze with ceramic centre
  • Compatible with Usbeck 4-feet stand
Fireproof stainless steel centre made of 1,4841 in format 60×60 mm, prevents burner flame penetration.
Weight: 40 g