Anti-KLF4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 56CT5.1.6]

Lieferant: US Biological
031072-PE
USBI031072-PEEA 884 EUR
USBI031072-PE
Anti-KLF4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 56CT5.1.6]
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:200-2000; Flow Cytometry: 1:10-50
Do not freeze! Light sensitive

Type: Primary
Antigen: KLF4 (Kruppel-like factor 4 (gut))
Clonality: Monoclonal
Clone: 56CT5.1.6
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR