Anti-TIMP1 Mouse Monoclonal Antibody [clone: 3F10F14]
Lieferant: US Biological
031291
USBI031291EA 649 EUR
USBI031291
Anti-TIMP1 Mouse Monoclonal Antibody [clone: 3F10F14]
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-250; Immunofluorescence: 1:10-50; Immunochemistry: 1:10-50
Type: Primary
Antigen: TIMP1 (TIMP metallopeptidase inhibitor 1)
Clonality: Monoclonal
Clone: 3F10F14
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human