Anti-TIMP1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 3F10F14]

Lieferant: US Biological
031291-HRP
USBI031291-HRPEA 884 EUR
USBI031291-HRP
Anti-TIMP1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 3F10F14]
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-250; Immunofluorescence: 1:10-50; Immunochemistry: 1:10-50

Type: Primary
Antigen: TIMP1 (TIMP metallopeptidase inhibitor 1)
Clonality: Monoclonal
Clone: 3F10F14
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR