Anti-AIF1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Lieferant: US Biological
031722-FITC
USBI031722-FITCEA 884 EUR
USBI031722-FITC
Anti-AIF1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100
Do not freeze!
Type: Primary
Antigen: AIF1 (allograft inflammatory factor 1)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse