Anti-EYA2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Lieferant: US Biological
035284-APC
USBI035284-APCEA 884 EUR
USBI035284-APC
Anti-EYA2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: EYA2 (eyes absent homolog 2 (Drosophila))
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human