Anti-KREMEN1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Lieferant: US Biological
037598-APC
USBI037598-APCEA 884 EUR
USBI037598-APC
Anti-KREMEN1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:50-100; Flow Cytometry: 1:10-50
Do not freeze!
Type: Primary
Antigen: KREMEN1 (kringle containing transmembrane protein 1)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human