Anti-SLC23A1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Lieferant: US Biological
041838-PE
USBI041838-PEEA 884 EUR
USBI041838-PE
Anti-SLC23A1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!
Type: Primary
Antigen: SLC23A1 (solute carrier family 23 (ascorbic acid transporter), member 1)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human