Anti-HPN Mouse Monoclonal Antibody [clone: 2D5]

Lieferant: US Biological
128060
USBI128060EA 623 EUR
USBI128060
Anti-HPN Mouse Monoclonal Antibody [clone: 2D5]
Antikörper
Type: Primary
Antigen: HPN (hepsin)
Clonality: monoclonal
Clone: 2D5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR