Anti-Isocitrate Dehydrogenase Mouse Polyclonal Antibody

Lieferant: US Biological
128614
USBI128614EA 623 EUR
USBI128614
Anti-Isocitrate Dehydrogenase Mouse Polyclonal Antibody
Antikörper
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: Isocitrate Dehydrogenase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR