Anti-ITGB1BP1 Mouse Monoclonal Antibody [clone: 3B2]

Lieferant: US Biological
128633
USBI128633EA 623 EUR
USBI128633
Anti-ITGB1BP1 Mouse Monoclonal Antibody [clone: 3B2]
Antikörper
Type: Primary
Antigen: ITGB1BP1 (integrin beta 1 binding protein 1)
Clonality: monoclonal
Clone: 3B2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR