Anti-MSRA Rabbit Polyclonal Antibody

Lieferant: US Biological
129936
USBI129936EA 662 EUR
USBI129936
Anti-MSRA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MSRA (methionine sulfoxide reductase A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR