Anti-TIMP1 Mouse Monoclonal Antibody [clone: 4D12]

Lieferant: US Biological
134455
USBI134455EA 623 EUR
USBI134455
Anti-TIMP1 Mouse Monoclonal Antibody [clone: 4D12]
Antikörper
Type: Primary
Antigen: TIMP1 (TIMP metallopeptidase inhibitor 1)
Clonality: monoclonal
Clone: 4D12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR