Anti-VAC14 Mouse Monoclonal Antibody [clone: 3B2]

Lieferant: US Biological
135195
USBI135195EA 623 EUR
USBI135195
Anti-VAC14 Mouse Monoclonal Antibody [clone: 3B2]
Antikörper
Recommended dilution: ELISA: 0.1ng/ml

Type: Primary
Antigen: VAC14 (Vac14 homolog (S. cerevisiae))
Clonality: monoclonal
Clone: 3B2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR